Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP39A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106814 Supplier Invitrogen™ Supplier No. PA5106814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome P450 39A1.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is involved in the conversion of cholesterol to bile acids. Its substrates include the oxysterols 25-hydroxycholesterol, 27-hydroxycholesterol and 24-hydroxycholesterol. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP39A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYP39A1
Gene Accession No. Q9JKJ9, Q9NYL5
Gene Alias 24-hydroxycholesterol 7-alpha-hydroxylase; Cyp39a1; Cytochrome P450 39A1; cytochrome P450 family 39 subfamily A member 1; cytochrome P450, 39a1 (oxysterol 7alpha-hydroxylase); cytochrome P450, family 39, subfamily A, polypeptide 1; cytochrome P450, subfamily XXXIX (oxysterol 7 alpha-hydroxylase), polypeptide 1; hCYP39A1; mCYP39A1; Oxysterol 7-alpha-hydroxylase
Gene Symbols CYP39A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYP39A1(Accession Q9NYL5), corresponding to amino acid residues L365-P415.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51302, 56050
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.