Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP39A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598970
Description
CYP39A1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is involved in the conversion of cholesterol to bile acids. Its substrates include the oxysterols 25-hydroxycholesterol, 27-hydroxycholesterol and 24-hydroxycholesterol. Alternate splicing results in multiple transcript variants.
Specifications
CYP39A1 | |
Polyclonal | |
Unconjugated | |
CYP39A1 | |
24-hydroxycholesterol 7-alpha-hydroxylase; Cyp39a1; Cytochrome P450 39A1; cytochrome P450 family 39 subfamily A member 1; cytochrome P450, 39a1 (oxysterol 7alpha-hydroxylase); cytochrome P450, family 39, subfamily A, polypeptide 1; cytochrome P450, subfamily XXXIX (oxysterol 7 alpha-hydroxylase), polypeptide 1; hCYP39A1; mCYP39A1; Oxysterol 7-alpha-hydroxylase | |
Rabbit | |
Protein G | |
RUO | |
51302 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.09 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9NYL5 | |
CYP39A1 | |
Recombinant Human 24-hydroxycholesterol 7-alpha-hydroxylase protein (235-392AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction