Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP3A4/CYP3A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101318
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101318 Supplier Invitrogen™ Supplier No. PA5101318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome P450 3A4/5.

CYP3A5 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. The enzyme metabolizes drugs such as nifedipine and cyclosporine as well as the steroid hormones testosterone, progesterone and androstenedione.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP3A4/CYP3A5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYP3A4
Gene Accession No. P08684, P20815
Gene Alias 1,8-cineole 2-exo-monooxygenase; albendazole monooxygenase; Albendazole monooxygenase (sulfoxide-forming); Albendazole sulfoxidase; aryl hydrocarbon hydroxylase; cholesterol 25-hydroxylase; CP33; CP34; CP35; CYP3A; CYP3A28; CYP3A3; CYP3A4; CYP3A5; CYPIIIA28; CYPIIIA3; CYPIIIA4; CYPIIIA5; cytochrome P450; cytochrome P450 3A28; cytochrome P450 3A3; Cytochrome P450 3A4; cytochrome P450 3A5; cytochrome P450 family 3 subfamily A member 4; cytochrome P450 family 3 subfamily A member 5; cytochrome P450 HLp; cytochrome P450 HLp2; cytochrome P450 NF-25; cytochrome P450, family 3, subfamily A, polypeptide 4; cytochrome P450, family 3, subfamily A, polypeptide 5; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5; cytochrome P450, subfamily IIIA, polypeptide 4; cytochrome P450-PCN1; Cytochrome P450-PCN3; flavoprotein-linked monooxygenase; glucocorticoid-inducible P450; HLP; HLp2; microsomal monooxygenase; NF-25; Nifedipine oxidase; P450C3; P450-III, steroid inducible; P450PCN1; P450PCN3; P450-PCN3; PCN3; Quinine 3-monooxygenase; taurochenodeoxycholate 6-alpha-hydroxylase; Unknown (protein for MGC:127840); xenobiotic monooxygenase
Gene Symbols CYP3A4, CYP3A5
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYP3A4(Accession P08684), corresponding to amino acid residues H267-I317.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1576, 1577
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.