Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP3A4 Recombinant Rabbit Monoclonal Antibody (10B10)
GREENER_CHOICE

Catalog No. FEMA549856
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549856 Supplier Invitrogen™ Supplier No. MA549856
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

CYP3A4 Recombinant Monoclonal Antibody for IHC (P), ELISA

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. This enzyme is involved in the metabolism of approximately half the drugs in use today, including acetaminophen, codeine, cyclosporin A, diazepam and erythromycin. The enzyme also metabolizes some steroids and carcinogens. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Previously another CYP3A gene, CYP3A3, was thought to exist; however, it is now thought that this sequence represents a transcript variant of CYP3A4. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP3A4
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 10B10
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYP3A4
Gene Accession No. P08684
Gene Alias 1,8-cineole 2-exo-monooxygenase; albendazole monooxygenase; Albendazole monooxygenase (sulfoxide-forming); Albendazole sulfoxidase; cholesterol 25-hydroxylase; CP33; CP34; CYP; CYP3A; CYP3A3; CYP3A4; CYPIIIA3; CYPIIIA4; Cytochrome P450 3A3; cytochrome P450 3A4; cytochrome P450 family 3 subfamily A member 4; cytochrome P450 HLp; Cytochrome P450 NF-25; cytochrome P450, family 3, subfamily A, polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; cytochrome P450-PCN1; glucocorticoid-inducible P450; HLP; MGC126680; NF-25; Nifedipine oxidase; P450; P450C3; P450-III, steroid inducible; P450PCN1; P450-PCN1; Quinine 3-monooxygenase; taurochenodeoxycholate 6-alpha-hydroxylase
Gene Symbols CYP3A4
Host Species Rabbit
Immunogen A synthesized peptide derived from human Cytochrome P450 3A4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1576
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.