Learn More
Invitrogen™ CYP3A5 Polyclonal Antibody

Description
CYP3A5 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF
Specifications
Specifications
| Antigen | CYP3A5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CYP3A5 |
| Gene Accession No. | P20815 |
| Gene Alias | aryl hydrocarbon hydroxylase; CP35; CYP3A28; CYP3A4; CYP3A5; CYPIIIA28; CYPIIIA5; cytochrome P450; cytochrome P450 3A28; cytochrome P450 3A5; cytochrome P450 family 3 subfamily A member 5; cytochrome P450 HLp2; cytochrome P450, family 3, subfamily A, polypeptide 5; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5; cytochrome P450-PCN3; flavoprotein-linked monooxygenase; HLp2; microsomal monooxygenase; P450PCN3; P450-PCN3; PCN3; Unknown (protein for MGC:127840); xenobiotic monooxygenase |
| Gene Symbols | CYP3A5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.