Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP3A7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$330.00 - $547.00
Specifications
Antigen | CYP3A7 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CYP3A7 Polyclonal antibody specifically detects CYP3A7 in Human samples. It is validated for Western BlotSpecifications
CYP3A7 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, pH 7.2, 40% glycerol | |
1551 | |
IgG | |
Affinity purified |
Western Blot 0.04-0.4 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
CYPIIIA7, cytochrome P450, family 3, subfamily A, polypeptide 7, Cytochrome P450-HFLA, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, subfamily IIIA, polypeptide 7, xenobiotic monooxygenase | |
This antibody was developed against Recombinant Protein corresponding to amino acids: LATHPDVQQKVQKEIDTVLPNKAPPTYD | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title