Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP3A7 Mouse anti-Human, Clone: F19 P2 H2, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $487.50
Specifications
Antigen | CYP3A7 |
---|---|
Clone | F19 P2 H2 |
Dilution | Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
CYP3A7 Monoclonal antibody specifically detects CYP3A7 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
CYP3A7 | |
Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CYPIIIA7, cytochrome P450, family 3, subfamily A, polypeptide 7, Cytochrome P450-HFLA, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, subfamily IIIA, polypeptide 7, xenobiotic monooxygenase | |
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminus of the human CYP3A7 protein and mouse myeloma Ag8563 cells. (Uniprot: Q9H241) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
F19 P2 H2 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Lipid and Metabolism | |
PBS | |
1551 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title