Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP3A7 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145345
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145345 200 μL
FEPA5145346 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145345 Supplier Invitrogen™ Supplier No. PA5145345
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene, CYP3A7, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme hydroxylates testosterone and dehydroepiandrosterone 3-sulphate, which is involved in the formation of estriol during pregnancy. The enzyme also metabolizes some drugs such as aflatoxin B1. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Transcript variants have been described, but it is not known whether these transcripts are normally produced.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP3A7
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene CYP3A7
Gene Accession No. P24462
Gene Alias aryl hydrocarbon hydroxylase; CP37; CYP3A7; CYPIIIA7; Cytochrome P450 3A7; cytochrome P450 family 3 subfamily A member 7; cytochrome P450, family 3, subfamily A, polypeptide 7; cytochrome P450, subfamily IIIA, polypeptide 7; cytochrome P450-HFLA; flavoprotein-linked monooxygenase; microsomal monooxygenase; P-450(HFL33); P-450111A7; P450-HFLA; P450HLp2; xenobiotic monooxygenase
Gene Symbols CYP3A7
Host Species Rabbit
Immunogen Recombinant Human CYP3A7 protein, Pro344-Asp497 (Accession #P24462).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1551
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.