Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP3C1 Antibody (V12P6F5*E3), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250210V
Description
CYP3C1 Monoclonal specifically detects CYP3C1 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CYP3C1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
V12P6F5*E3 | |
Western Blot, ELISA, Immunohistochemistry | |
CYP3A, cyp3c1, Cytochrome P450, family 3, subfamily c, polypeptide 1, wu:fa96d12, wu:fc26b04, zgc:56626 | |
Ovalbumin-conjugated synthetic peptide KLIPRSHKEK (C-terminal sequence) | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction