Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP46A1 Antibody (N8-P6E4*H8), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250194AF750
Description
CYP46A1 Monoclonal specifically detects CYP46A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CYP46A1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CYP46A1 | |
Ovalbumin-conjugated synthetic peptide- PVLCTLRPR (C-terminal sequence) | |
0.1 mL | |
10858 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry | |
N8-P6E4*H8 | |
Western Blot, ELISA, Immunohistochemistry | |
CH24H, cholesterol 24-hydroxylase, CYP46CP46, Cytochrome P450 46A1, cytochrome P450, family 46, subfamily A, polypeptide 1, cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase), EC 1.14.13.98 | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction