Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP46A1 Antibody (N8-P6E4*H8), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250194MFV500
Description
CYP46A1 Monoclonal antibody specifically detects CYP46A1 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| CYP46A1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| N8-P6E4*H8 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CH24H, cholesterol 24-hydroxylase, CYP46CP46, Cytochrome P450 46A1, cytochrome P450, family 46, subfamily A, polypeptide 1, cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase), EC 1.14.13.98 | |
| Ovalbumin-conjugated synthetic peptide- PVLCTLRPR (C-terminal sequence) | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience | |
| 10858 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction