Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP46A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$181.90 - $487.50
Specifications
Antigen | CYP46A1 |
---|---|
Dilution | Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400140SS
![]() |
Novus Biologicals
NB400140SS |
0.025 mL |
Each for $181.90
|
|
|||||
NB400140
![]() |
Novus Biologicals
NB400140 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
CYP46A1 Polyclonal specifically detects CYP46A1 in Human, Rabbit, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CYP46A1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Neuroscience | |
Whole antisera with No Preservative | |
CH24H, cholesterol 24-hydroxylase, CYP46CP46, Cytochrome P450 46A1, cytochrome P450, family 46, subfamily A, polypeptide 1, cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase), EC 1.14.13.98 | |
CYP46A1 | |
IgG | |
Unpurified | |
Human and Rabbit. Does not react with mouse. |
Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Rabbit, Mouse (Negative) | |
Q9Y6A2 | |
10858 | |
A synthetic peptide corresponding to residues between 200-250 of human CYP46A1. [UniProt# Q9Y6A2] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title