Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4A11/22 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986304
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6304 Supplier Bioss Supplier No. BS5055R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and hydroxylates medium-chain fatty acids such as laurate and myristate.

Specifications

Antigen CYP4A11/22
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CYP4A11
Gene Accession No. Q02928
Gene Alias 20-HETE synthase; 20-hydroxyeicosatetraenoic acid synthase; alkane-1 monooxygenase; CP4Y; CYP4A11; CYP4A2; Cyp4a-2; CYP4AII; CYPIVA11; CYPIVA2; Cytochrome P450 4A11; Cytochrome P450 4A2; cytochrome P450 family 4 subfamily A member 11; Cytochrome P-450 K-2; Cytochrome P450 K-5; cytochrome P450, family 4, subfamily A, polypeptide 11; cytochrome P450, family 4, subfamily a, polypeptide 2; cytochrome P450, subfamily 4A, polypeptide 11; cytochrome P450, subfamily IVA, polypeptide 11; Cytochrome P-450HK-omega; Cytochrome P450HL-omega; Cytochrome P450-LA-omega 2; Fatty acid omega-hydroxylase; Lauric acid omega-hydroxylase; Long-chain fatty acid omega-monooxygenase; P-450 HK omega; P-450 IV4AII; P-450 K-2; P450 K-5; P450-HL-omega; P450HL-omega; P450-LA-omega 2
Gene Symbols CYP4A11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CYP4A22.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1579
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.