Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4A11/CYP4A22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102018 Supplier Invitrogen™ Supplier No. PA5102018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome P450 4A11/22.

Cytochrome P450 4A22 (CYP4A22) is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 1p33.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4A11/CYP4A22
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYP4A22
Gene Accession No. Q02928, Q5TCH4
Gene Alias 20-HETE synthase; 20-hydroxyeicosatetraenoic acid synthase; alkane-1 monooxygenase; CP4Y; Cyp4a1; Cyp4a-1; Cyp4a10; Cyp4a11; CYP4A2; Cyp4a-2; CYP4A22; CYP4AII; CYPIVA10; CYPIVA11; CYPIVA2; CYPIVA22; Cytochrome P450 4A10; cytochrome P450 4A11; Cytochrome P450 4A2; cytochrome P450 4A22; cytochrome P450 4A22K; cytochrome P450 family 4 subfamily A member 11; cytochrome P450 family 4 subfamily A member 22; Cytochrome P-450 K-2; cytochrome P450 K-5; cytochrome P450, 4A1; cytochrome P450, 4a10; cytochrome P450, family 4, subfamily a, polypeptide 1; cytochrome P450, family 4, subfamily a, polypeptide 10; cytochrome P450, family 4, subfamily A, polypeptide 11; cytochrome P450, family 4, subfamily a, polypeptide 2; cytochrome P450, family 4, subfamily A, polypeptide 22; cytochrome P450, IVA1; cytochrome P450, subfamily 4A, polypeptide 11; cytochrome P450, subfamily IVA, polypeptide 11; cytochrome P-450HK-omega; cytochrome P450HL-omega; cytochrome P450-LA-omega 1; Cytochrome P450-LA-omega 2; Cytochrome P452; fatty acid omega-hydroxylase; Lauric acid omega-hydroxylase; long-chain fatty acid omega-monooxygenase; P-450 IV4AII; P-450 K-2; P450 K-5; P450HL-omega; P450-LA-omega 2
Gene Symbols CYP4A11, CYP4A22
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYP4A11(Accession Q02928), corresponding to amino acid residues F150-C200.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1579, 284541
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.