Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521497 Supplier Invitrogen™ Supplier No. PA521497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and hydroxylates medium-chain fatty acids such as laurate and myristate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4A11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CYP4A11
Gene Accession No. Q02928
Gene Alias 20-HETE synthase; 20-hydroxyeicosatetraenoic acid synthase; alkane-1 monooxygenase; CP4Y; Cyp4a11; Cyp4a2; Cyp4a-2; CYP4AII; CYPIVA11; CYPIVA2; cytochrome P450 4A11; Cytochrome P450 4A2; cytochrome P450 family 4 subfamily A member 11; cytochrome P-450 K-2; cytochrome P450 K-5; cytochrome P450, family 4, subfamily A, polypeptide 11; cytochrome P450, family 4, subfamily a, polypeptide 2; cytochrome P450, subfamily 4A, polypeptide 11; cytochrome P450, subfamily IVA, polypeptide 11; Cytochrome P-450HK-omega; Cytochrome P450HL-omega; cytochrome P450-LA-omega 2; fatty acid omega-hydroxylase; Lauric acid omega-hydroxylase; long-chain fatty acid omega-monooxygenase; P-450 HK omega; P-450 IV4AII; P-450 K-2; P450 K-5; P450-HL-omega; P450HL-omega; P450-LA-omega 2
Gene Symbols CYP4A11
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CYP4A11. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1579
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.