Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4A22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113742 Supplier Invitrogen™ Supplier No. PA5113742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQLYLHRQWL LKALQQFPCP PSHWLFGHIQ EFQHDQELQR IQERVKTFPS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

CYP4A22 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids.This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 1p33.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4A22
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CYP4A22
Gene Accession No. Q5TCH4
Gene Alias CYP4A22; CYPIVA22; cytochrome P450 4A22; cytochrome P450 4A22K; cytochrome P450 family 4 subfamily A member 22; cytochrome P450, family 4, subfamily A, polypeptide 22; fatty acid omega-hydroxylase; Lauric acid omega-hydroxylase; long-chain fatty acid omega-monooxygenase; RP1-18D14.1
Gene Symbols CYP4A22
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human CYP4A22 (aa 35-84).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284541
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.