Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582148 Supplier Invitrogen™ Supplier No. PA582148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGSLDKVVSW AHQFPYAHPL WFGQFIGFLN IYEPDYAKAV YSRGDPKAPD VYDFFLQWIG RGLLVLEGPK WLQHRKLLTP GFHYDVLKPY VAVFTESTRI MLDKWEEKAR EGKSFDIFCD VGH Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 87%.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. In rodents, the homologous protein has been shown to metabolize certain carcinogens; however, the specific function of the human protein has not been determined. Two transcript variants encoding slightly different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4B1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYP4B1
Gene Accession No. P13584
Gene Alias CYP4B1; Cyp4b-1; CYP4B1 isoform; CYP4B1-like isozyme short form; CYPIVB1; cytochrome P450 4B1; cytochrome P450 family 4 subfamily B member 1; cytochrome P450 isozyme 5; cytochrome P-450 isozyme 5; Cytochrome P450 L-2; cytochrome P450, family 4, subfamily B, polypeptide 1; cytochrome P450, subfamily 4B, polypeptide 1; cytochrome P450, subfamily IV B, polypeptide 1; cytochrome P450, subfamily IVB, polypeptide 1; cytochrome P450-HP; microsomal monooxygenase; P450 L-2; P-450HP; P450-isozyme 5
Gene Symbols CYP4B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYP4B1. Recombinant protein control fragment (Product #RP-101024).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1580
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.