Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4F11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553570 Supplier Invitrogen™ Supplier No. PA553570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDNCRRLQCF PQPPKQNWFW GHQGLVTPTE EGMKTLTQLV TTYPQGFKLW LGPTFPLLIL CHPDIIRPIT SASAAVAPKD MIFYGFLKPW LGDGLLLSGG DKWSRHRRML TPAF Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.

The cytochrome P450 proteins (CYPs) are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. P450 enzymes are classified into subfamilies based on their sequence similarities. CYP4F11 is an isoform of the cytochrome P450 4F protein and is expressed mainly in human liver, followed by kidney, heart, and skeletal muscle. CYP4F11 contains 524 amino acid residues that share 80.0, 82.3 and 79.2% identity to CYP4F2, CYP4F3, and CYP4F8 amino acid sequences, respectively. CYP4F11 has catalytic properties towards endogenous eicosanoids as well as some clinically relevant drugs, and is able to metabolize large molecules such as erythromycin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4F11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYP4F11
Gene Accession No. Q9HBI6
Gene Alias 3-hydroxy fatty acids omega-hydroxylase CYP4F11; CYP4F11; CYPIVF11; cytochrome P450 4F11; cytochrome P450 family 4 subfamily F member 11; cytochrome P450, family 4, subfamily F, polypeptide 11; cytochrome P450, subfamily IVF, polypeptide 11; Docosahexaenoic acid omega-hydroxylase; Long-chain fatty acid omega-monooxygenase; Phylloquinone omega-hydroxylase CYP4F11
Gene Symbols CYP4F11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYP4F11. Recombinant protein control fragment (Product #RP-90473).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.