Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4F12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596890
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596890 Supplier Invitrogen™ Supplier No. PA596890
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DFTDAVIRER RRTLPTQGID DFFKDKAKSK TLDFIDVLLL SKDEDGKALS DEDIRAEADT FMFGGHDTTA SGLSWVLYNL ARHPEYQERC RQEVQELLKD RDPKEIEWDD LAQLPFLTMC VKESLRLHPP APFISRCCTQ DIVLPDGRVI PKGITCLIDI IGVHHNPTVW PDPEVYDPFR FDPENSKGRS PLAFIPFSAG PRNCIGQAFA MAEMKVVLAL MLLHFRFLPD HTEPRRKLEL IMRAEGGLWL RVEPLNVSLQ; Positive Samples: Mouse placenta, Rat brain; Cellular Location: Endoplasmic reticulum membrane, Microsome membrane, Multi-pass membrane protein.

CYP4F12 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein likely localizes to the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4F12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYP4F12
Gene Accession No. Q9HCS2
Gene Alias CYP4F12; CYPIVF12; Cytochrome P450 4F12; cytochrome P450 family 4 subfamily F member 12; cytochrome P450, family 4, subfamily F, polypeptide 12; cytochrome P450, subfamily IVF, polypeptide 12; F22329_1; UNQ568/PRO1129
Gene Symbols CYP4F12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 265-524 of human CYP4F12 (NP_0764333).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66002
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.