Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP4F2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216070
Description
CYP4F2 Polyclonal specifically detects CYP4F2 in Human, Porcine samples. It is validated for Western Blot.Specifications
CYP4F2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
CPF2, CYPIVF2, Cytochrome P450 4F2, cytochrome P450, family 4, subfamily F, polypeptide 2, cytochrome P450, subfamily IVF, polypeptide 2, Cytochrome P450-LTB-omega, EC 1.14.13.30, leukotriene B4 omega-hydroxylase, Leukotriene-B(4) 20-monooxygenase 1, leukotriene-B(4) omega-hydroxylase 1, leukotriene-B4 20-monooxygenase | |
Rabbit | |
60 kDa | |
0.1 mL | |
Lipid and Metabolism | |
8529 | |
Human, Pig | |
IgG |
Western Blot | |
0.28mg/mL | |
Western Blot 1:500-1:3000 | |
P78329 | |
CYP4F2 | |
Recombinant protein encompassing a sequence within the center region of human Cytochrome P450 4F2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Bovine (87%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction