Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4F2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587655 Supplier Invitrogen™ Supplier No. PA587655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFYDNCRRLR CFPQPPRRNW FWGHQGMVNP TEEGMRVLTQ LVATYPQGFK VWMGPISPLL SLCHPDIIRS VINASAAIAP K; Positive Samples: LO2, HepG2, 293T; Cellular Location: Endoplasmic reticulum membrane, Microsome membrane, Peripheral membrane protein.

CYP4F isoforms are involved in the oxidation of important cellular mediators, such as leukotriene B4 (LTB4) and prostaglandins. CYP4F2 catalyzes omega-hydroxylation of LTB4 to a less potent proinflammatory eicosanoid, 20-OH-LTB4 as well as arachidonic acid. CYP4F2 is expressed in liver and kidney. Its expression is repressed by peroxisomal proliferators and induced by retinoic acid. X-linked adrenoleukodystrophy (X-ALD) is a severe neurodegenerative disorder biochemically characterized by elevated levels of very long-chain fatty acids (VLCFA). CYP4F2 participates in the omega-hydroxylation of VLCFAs, which may provide an alternative mode of treatment for X-ALD patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4F2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYP4F2
Gene Accession No. P78329
Gene Alias 20-HETE synthase; 20-hydroxyeicosatetraenoic acid synthase; Arachidonic acid omega-hydroxylase; CPF2; CYP4F2; CYPIVF2; cytochrome P450 4F2; cytochrome P450 family 4 subfamily F member 2; cytochrome P450, family 4, subfamily F, polypeptide 2; cytochrome P450, subfamily IVF, polypeptide 2; cytochrome P450-LTB-omega; Docosahexaenoic acid omega-hydroxylase; docosahexaenoic acid omega-hydroxylase CYP4F3; phylloquinone omega-hydroxylase CYP4F2; leukotriene-B(4) 20-monooxygenase 1; Leukotriene-B(4) omega-hydroxylase 1; phylloquinone omega-hydroxylase CYP4F2
Gene Symbols CYP4F2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human CYP4F2 (NP_0010733).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8529
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.