Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP4F22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568022
Description
CYP4F22 Polyclonal Antibody for Western Blot
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 19 and encodes an enzyme thought to play a role in the 12(R)-lipoxygenase pathway. Mutations in this gene are the cause of ichthyosis lamellar type 3.
Specifications
CYP4F22 | |
Polyclonal | |
Unconjugated | |
CYP4F22 | |
ARCI5; CYP4F22; Cytochrome P450 4F22; cytochrome P450 family 4 subfamily F member 22; cytochrome P450, family 2, subfamily E, polypeptide 2 homolog; cytochrome P450, family 4, subfamily F, polypeptide 22; INLNE; LI3 | |
Rabbit | |
affinity chromatography | |
RUO | |
126410 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6NT55 | |
CYP4F22 | |
Synthesized peptide derived from human CYP4F22. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction