Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP4F3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169678
Description
CYP4F3 Polyclonal specifically detects CYP4F3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CYP4F3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CYP4F, CYPIVF3, cytochrome P-450, Cytochrome P450 4F3, cytochrome P450, family 4, subfamily F, polypeptide 3, cytochrome P450, subfamily IVF, polypeptide 3 (leukotriene B4 omegahydroxylase), Cytochrome P450-LTB-omega, EC 1.14.13.30, leukotriene B4 omega hydroxylase, Leukotriene-B(4) 20-monooxygenase 2, leukotriene-B(4) omega-hydroxylase 2, leukotriene-B4 20-monooxygenase, LTB4HCPF3 | |
Rabbit | |
60 kDa | |
100 μL | |
Lipid and Metabolism | |
4051 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin | |
Q08477 | |
CYP4F3 | |
Synthetic peptides corresponding to CYP4F3(cytochrome P450, family 4, subfamily F, polypeptide 3) The peptide sequence was selected from the N terminal of CYP4F3. Peptide sequence LAWTYTFYDNCCRLRCFPQPPKRNWFLGHLGLIHSSEEGLLYTQSLACTF. | |
Affinity purified | |
RUO | |
Primary | |
Canine: 86%; Rat: 85%; Mouse: 85%. | |
Human, Mouse, Rat, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction