Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4F8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA515057
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA515057 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA515057 Supplier Invitrogen™ Supplier No. PA515057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP4F8 Polyclonal Antibody for Western Blot

CYP4F8 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and functions as a 19-hydroxylase of prostaglandins in seminal vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4F8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CYP4F8
Gene Accession No. P98187
Gene Alias CPF8; CYP4F8; CYPIVF8; Cytochrome P450 4F8; cytochrome P450 family 4 subfamily F member 8; cytochrome P450, family 4, subfamily F, polypeptide 8; cytochrome P450, subfamily IVF, polypeptide 8; flavoprotein-linked monooxygenase; microsomal monooxygenase
Gene Symbols CYP4F8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 473-503 amino acids from the C-terminal region of human CYP4F8.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11283
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.