Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP4V2 Antibody (M29-P3B10), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250214MFV500
Description
CYP4V2 Monoclonal antibody specifically detects CYP4V2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| CYP4V2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| M29-P3B10 | |
| Western Blot, ELISA, Immunohistochemistry | |
| BCD, CYP4AH1, cytochrome P450 4V2, cytochrome P450, family 4, subfamily V, polypeptide 2, EC 1.14, EC 1.14.14.1, FLJ18432, MGC43534 | |
| Peptide sequence - KREELGLEGQ (amino acids 495 - 504) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 285440 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction