Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4X1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen CYP4X1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP162382
SDP
View Documents
Novus Biologicals
NBP162382
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

CYP4X1 Polyclonal specifically detects CYP4X1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CYP4X1
Polyclonal
Rabbit
Human
CYPIVX1, cytochrome P450 4X1, cytochrome P450, family 4, subfamily X, polypeptide 1, EC 1.14.14.1, MGC40051
CYP4X1
IgG
59 kDa
Western Blot
Unconjugated
RUO
Q8N118
260293
Synthetic peptides corresponding to CYP4X1(cytochrome P450, family 4, subfamily X, polypeptide 1) The peptide sequence was selected from the middle region of CYP4X1. Peptide sequence LDIVLSAKDESGSSFSDIDVHSEVSTFLLAGHDTLAASISWILYCLALNP.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.