Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP4X1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115087
Description
Antibody detects endogenous levels of total Cytochrome P450 4X1.
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma. The CYP4X1 gene product has been provisionally designated CYP4X1 pending further characterization.
Specifications
CYP4X1 | |
Polyclonal | |
Unconjugated | |
CYP4X1 | |
A230025G20; CP4X1; CYP_a; Cyp4a28-ps; Cyp4x1; CYPIVX1; cytochrome P450 4X1; cytochrome P450 family 4 subfamily X member 1; cytochrome P450, 4x1; cytochrome P450, family 4, subfamily X, polypeptide 1; EC 1.14.14.1; PRO4404; UNQ1929; UNQ1929/PRO4404 | |
Rabbit | |
Affinity chromatography | |
RUO | |
260293 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q8N118 | |
CYP4X1 | |
A synthesized peptide derived from human CYP4X1(Accession Q8N118), corresponding to amino acid residues V253-D303. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction