Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    CYP4Z1 Antibody (N7-P2G5*D8), mFluor Violet 450 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250215MFV450
Description
CYP4Z1 Monoclonal antibody specifically detects CYP4Z1 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| CYP4Z1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| Western Blot, ELISA, Immunohistochemistry | |
| N7-P2G5*D8 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CYP4A20, CYPIVZ1, cytochrome P450 4Z1, cytochrome P450, family 4, subfamily Z, polypeptide 1, EC 1.14.14.1 | |
| Ovalbumin-conjugated synthetic peptide KLAPDHSRPP (C-terminal sequence) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cell Cycle and Replication, Cellular Signaling, metabolism | |
| 199974 | |
| Store at 4C in the dark. | |
| IgG1 κ | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            