Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4Z2P Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989782
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9782 Supplier Bioss Supplier No. BS14164R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The cytochrome P450 (P450) superfamily of enzymes is one of three enzyme systems which metabolize the fatty acid arachadonic acid (AA) to regulators of vascular tone. P450 enzymes are monooxygenase enzymes which require several co-factors such as nicotinamide adenine dinucleotide phosphate (NADPH) and P450 reductase. There are over 200 known genes which encode P450s. Epoxygenases are those P450s which metabolize AA to epoxyeicosatrienoic acid (EETs) and omega-hydroxylases are those P450s which produce 19- and 20-hydroxyeicosatetraenoic acids (19- and 20-HETE). As well as fatty acid metabolism, P450s also metabolize many drugs and toxins. Cytochrome P450 3A4 is abundantly expressed in liver and small intestine and is inducible by barbiturates, glucocorticoids and rifampicin.

Specifications

Antigen CYP4Z2P
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene CYP4Z2P
Gene Accession No. Q86W10, Q8N1L4
Gene Alias CYP4A20; CYP4Z1; CYP4Z1/2; CYP4Z2P; CYPIVZ1; Cytochrome P450 4Z1; cytochrome P450 family 4 subfamily Z member 1; cytochrome P450 family 4 subfamily Z member 2, pseudogene; cytochrome P450, family 4, subfamily Z, polypeptide 1; P450 4Z1/2; UNQ3060/PRO9882
Gene Symbols CYP4Z1, CYP4Z2P
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CYP4Z2P.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163720, 199974
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.