Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP51A1 Antibody (N6-P2H5*G8), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250216AF750
Description
CYP51A1 Monoclonal specifically detects CYP51A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CYP51A1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CYP51A1 | |
Ovalbumin-conjugated synthetic peptide NPVIRYKRRS (C-terminal sequence). | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
N6-P2H5*G8 | |
Western Blot, ELISA, Immunohistochemistry | |
CP51, CYP51Cytochrome P450LI, CYPL1, CYPLI, Cytochrome P450 51A1, cytochrome P450, family 51, subfamily A, polypeptide 1, Cytochrome P450-14DM, EC 1.14.13.70, lanosterol 14-alpha demethylase, lanosterol 14-alpha-demethylase, LDMCytochrome P45014DM, P450-14DM, P450L1,51 (lanosterol 14-alpha-demethylase), Sterol 14-alpha demethylase | |
Mouse | |
Protein A purified | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
1595.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction