Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CYP51A1, Mouse, Unlabeled, Clone: N6-P2H5*G8,

Catalog No. MABS1259MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1259MI 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1259MI Supplier MilliporeSigma™ Supplier No. MABS1259
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lanosterol 14-alpha demethylase (EC 1.14.13.70; UniProt Q16850; also known as CYP51A1, CYPL1, CYPLI, Cytochrome P450 51A1, Cytochrome P450, family 51, subfamily A, polypeptide 1, Cytochrome P450-14DM, Cytochrome P45014DM, Cytochrome P450L1, Cytochrome P450LI, LDM, Sterol 14-alpha demethylase) is encoded by the CYP51A1 (also known as CYP51) gene (Gene ID 1595) in human. Cytochromes P450 (CYP) proteins are primarily membrane-associated oxidases located either in the inner membrane of mitochondria or in the endoplasmic reticulum where they function as the terminal enzymes in electron transfer chains. In addition to processing endogenous substrates, CYPs also function to metabolize exogenous drugs and potentially toxic chemicals. The human CYP superfamily consisits of 57 genes and more than 59 pseudogenes divided into 18 families and 43 subfamilies. CYP51A1 represents the only member of the CYP51 subfamily and is involved in the conversion of lanosterol to 4,4-dimethylcholesta-8(9),14,24-trien-3 -ol. Products generated by the CYP51 reaction are vital intermediates in pathways leading to the formation of cholesterol in humans, ergosterol in fungi, and other types of sterols in plants. Sterols play an important structural role in the regulation of membrane fluidity and permeability, which influence the functions of enzymes, ion channels, and other cell components embedded within.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP51A1
Applications Western Blot
Classification Monoclonal
Clone N6-P2H5*G8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene CYP51A1, CYP51
Gene Accession No. Q16850
Host Species Mouse
Immunogen Ovalbumin-conjugated linear peptide corresponging to a sequence near the C-terminus of human/mouse/rat CYP51A1.
Purification Method Protein G Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000777
Test Specificity Clone N6-P2H5*G8 was raised against a CYP51A1 C-terminal sequence 100% conserved among human, mouse, and rat species. Target band was detected only in lysate from CYP51A1-transfected, but not from untransfected, cells (Swan, R., et al. (2016). Oncotarget. In press).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.