Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP51A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589258 Supplier Invitrogen™ Supplier No. PA589258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WLLPGWLPLP SFRRRDRAHR EIKDIFYKAI QKRRQSQEKI DDILQTLLDA TYKDGRPLTD DEVAGMLIGL LLAGQHTSST TSAWMGFFLA RDKTLQKKCY LEQKTVCGEN LPPLTYDQLK DLNLLDRCIK ETLRLRPPIM IMMRMARTPQ TVAGYTIPPG HQVCVSPTVN QRLKDS Positive Samples: Cellular Location: Endoplasmic reticulum membrane, Microsome membrane.

P450 enzymes constitute a family of monooxygenase enzymes that are involved in the metabolism of a wide array of endogenous and xenobiotic compounds. Several P450 enzymes have been classified by sequence similarities as members of the CYP1A and CYP2A subfamilies. CYP51A1 (cytochrome P450, family 51, subfamily A, polypeptide 1), also known as LDM (lanosterol 14-alpha demethylase) or cytochrome P450-14DM, is a 503 amino acid protein localized to endoplasmic reticulum membrane. CYP51A1 is an important enzyme for zymosterol and steroid biosynthesis. CYP51A1 catalyzes C14-demethylation of lanosterol, transforming lanosterol into 4,4'-dimethyl cholesta-8,14,24-triene-3-beta-ol. CYP51A1 is ubiquitously expressed, with highest levels found in liver, ovary, testis, lung, kidney and prostate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP51A1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene CYP51A1
Gene Accession No. Q16850, Q64654, Q8K0C4
Gene Alias AI426508; CP51; CYP51; CYP51A1; CYPL1; CYPLI; Cytochrom P450 Lanosterol 14 alpha-demethylase; Cytochrome P450 51; cytochrome P450 51A1; cytochrome P450 family 51 subfamily A member 1; cytochrome P450 Lanosterol 14 alpha-demethylase; cytochrome P450, 51; cytochrome P450, 51 (lanosterol 14-alpha-demethylase); cytochrome P450, family 51; cytochrome P450, family 51, subfamily A, polypeptide 1; cytochrome P450, subfamily 51; Cytochrome P45014DM; cytochrome P450-14DM; cytochrome P450LI; Lanosterol 14-alpha demethylase; lanosterol 14-alpha-demethylase; LDM; P450 14DM; P45014DM; P450-14DM; P450L1; P450LI; RATCP14DM; Sterol 14-alpha demethylase
Gene Symbols Cyp51, CYP51A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 245-420 of CYP51A1 (NP_0007771).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13121, 1595, 25427
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.