Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP51A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598400
Description
CYP51A1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
P450 enzymes constitute a family of monooxygenase enzymes that are involved in the metabolism of a wide array of endogenous and xenobiotic compounds. Several P450 enzymes have been classified by sequence similarities as members of the CYP1A and CYP2A subfamilies. CYP51A1 (cytochrome P450, family 51, subfamily A, polypeptide 1), also known as LDM (lanosterol 14-alpha demethylase) or cytochrome P450-14DM, is a 503 amino acid protein localized to endoplasmic reticulum membrane. CYP51A1 is an important enzyme for zymosterol and steroid biosynthesis. CYP51A1 catalyzes C14-demethylation of lanosterol, transforming lanosterol into 4,4'-dimethyl cholesta-8,14,24-triene-3-beta-ol. CYP51A1 is ubiquitously expressed, with highest levels found in liver, ovary, testis, lung, kidney and prostate.
Specifications
CYP51A1 | |
Polyclonal | |
Unconjugated | |
CYP51A1 | |
AI426508; CP51; CYP51; CYP51A1; CYPL1; CYPLI; Cytochrom P450 Lanosterol 14 alpha-demethylase; Cytochrome P450 51; cytochrome P450 51A1; cytochrome P450 family 51 subfamily A member 1; cytochrome P450 Lanosterol 14 alpha-demethylase; cytochrome P450, 51; cytochrome P450, 51 (lanosterol 14-alpha-demethylase); cytochrome P450, family 51; cytochrome P450, family 51, subfamily A, polypeptide 1; cytochrome P450, subfamily 51; Cytochrome P45014DM; cytochrome P450-14DM; cytochrome P450LI; Lanosterol 14-alpha demethylase; lanosterol 14-alpha-demethylase; LDM; P450 14DM; P45014DM; P450-14DM; P450L1; P450LI; RATCP14DM; Sterol 14-alpha demethylase | |
Rabbit | |
Protein G | |
RUO | |
1595 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q16850 | |
CYP51A1 | |
Recombinant Human Lanosterol 14-alpha demethylase protein (270-437AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction