Learn More
Invitrogen™ CYP7A1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total Cytochrome P450 7A1.
Specifications
Specifications
| Antigen | CYP7A1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Cyp7a1 |
| Gene Accession No. | P18125, P22680, Q64505 |
| Gene Alias | 24-hydroxycholesterol 7-alpha-hydroxylase; CHAP; cholesterol 7 alpha hydroxylase; cholesterol 7-alpha hydroxylase; cholesterol 7-alpha-hydroxylase; Cholesterol 7-alpha-monooxygenase; CP7A; CYP7; Cyp7a1; CYP7S1; CYPVII; cytochrome P450 (cholesterol hydroxylase 7 alpha); cytochrome P450 7A1; cytochrome P450 family 7 subfamily A member 1; cytochrome P450, 7a1; cytochrome P450, family 7, subfamily a, polypeptide 1; cytochrome P450, subfamily VIIA polypeptide 1 |
| Gene Symbols | Cyp7a1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.