Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP7B1 Antibody (M17-P3F2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250217R
Description
CYP7B1 Monoclonal specifically detects CYP7B1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CYP7B1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYP7B1 | |
Ovalbumin-conjugated synthetic peptide IQYPDSDVL (C-terminal sequence) | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience | |
9420 | |
Store at 4C in the dark. | |
IgG1 λ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
M17-P3F2 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CBAS3, CP7B, Cytochrome P450 7B1, cytochrome P450, family 7, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1,25-hydroxycholesterol 7-alpha-hydroxylase, EC 1.14.13.100, oxysterol 7alpha-hydroxylase, Oxysterol 7-alpha-hydroxylase, spastic paraplegia 5A (autosomal recessive), SPG5A | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction