Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP7B1 Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
SDP

Catalog No. 502544080
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4080 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4080 Supplier OriGene Supplier No. CF807549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP7B1 Monoclonal antibody specifically detects CYP7B1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CYP7B1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1G7
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene DMRT1
Gene Accession No. Q9Y5R6
Gene Alias 24-hydroxycholesterol 7-alpha-hydroxylase; 25/26-hydroxycholesterol 7-alpha-hydroxylase; 25-hydroxycholesterol 7-alpha-hydroxylase; 3-hydroxysteroid 7-alpha hydroxylase; AW261589; CBAS3; CP7B; CYP7B1; Cytochrome P450 7B1; cytochrome P450 family 7 subfamily B member 1; cytochrome P450, 7b1; cytochrome P450, family 7, subfamily b, polypeptide 1; cytochrome P450, subfamily 7B, polypeptide 1; cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1; D3Ertd552e; hct-1; hippocampal transcript 1 protein; Oxysterol 7-alpha-hydroxylase; SPG5A
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-210 of human DMRT1 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1761
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.