Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP7B1 Monoclonal Antibody (OTI7H10)
GREENER_CHOICE

Catalog No. PIMA524898
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524898 Supplier Invitrogen™ Supplier No. MA524898
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP7B1 Monoclonal Antibody for Western Blot

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum membrane protein catalyzes the first reaction in the cholesterol catabolic pathway of extrahepatic tissues, which converts cholesterol to bile acids. This enzyme likely plays a minor role in total bile acid synthesis, but may also be involved in the development of atherosclerosis, neurosteroid metabolism and sex hormone synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP7B1
Applications Western Blot
Classification Monoclonal
Clone OTI7H10
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cyp7b1
Gene Accession No. O75881
Gene Alias 24-hydroxycholesterol 7-alpha-hydroxylase; 25/26-hydroxycholesterol 7-alpha-hydroxylase; 25-hydroxycholesterol 7-alpha-hydroxylase; 3-hydroxysteroid 7-alpha hydroxylase; AW261589; CBAS3; CP7B; CYP7B1; Cytochrome P450 7B1; cytochrome P450 family 7 subfamily B member 1; cytochrome P450, 7b1; cytochrome P450, family 7, subfamily b, polypeptide 1; cytochrome P450, subfamily 7B, polypeptide 1; cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1; D3Ertd552e; hct-1; hippocampal transcript 1 protein; Oxysterol 7-alpha-hydroxylase; SPG5A
Gene Symbols Cyp7b1
Host Species Mouse
Immunogen Recombinant protein expressed in E.coli corresponding to amino acids 350-506 of Cyp7b1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9420
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.