Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP8B1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP168884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP168884 100 μL
NBP16888420 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP168884 Supplier Novus Biologicals Supplier No. NBP168884
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP8B1 Polyclonal specifically detects CYP8B1 in Human samples. It is validated for Western Blot.

Specifications

Antigen CYP8B1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UNU6
Gene Alias CP8B, CYP127 alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase, CYPVIIIB1, Cytochrome P450 8B1, cytochrome P450, family 8, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIIB (sterol 12-alpha-hydroxylase), polypeptide 1,7-alpha-hydroxycholest-4-en-3-one 12-alpha-hydroxylase, EC 1.14.13.95, FLJ17826,7-alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase, Sterol 12-alpha-hydroxylase
Gene Symbols CYP8B1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CYP8B1 (cytochrome P450, family 8, subfamily B, polypeptide 1) The peptide sequence was selected from the middle region of CYP8B1. Peptide sequence SLLWPREWLEVGRLQRLFHKMLSVSHSQEKEGISNWLGNMLQFLREQGVP.
Molecular Weight of Antigen 58 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1582
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.