Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP8B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102021 Supplier Invitrogen™ Supplier No. PA5102021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome P450 8B1.

CYP8B1 (sterol 12-alpha-hydroxylase) is a member of the cytochrome P450 superfamily of monooxygenase enzymes that are involved in the metabolism of a wide array of endogenous and xenobiotic compounds. CYP8B1 is highly expressed in liver and is an important enzyme for bile acid synthesis. Specifically, CYP8B1 moderates the ratio of cholic acid over chenodeoxycholic acid to control the solubility of cholesterol. The gene encoding human CYP8B1 maps to chromosome 3p22.1. The CYP8B1 gene encodes a 501-amino acid protein and does not contain any introns. The CYP8B1 gene promoter is transactivated by hepatocyte nuclear factor 4a. In mice, disruption of the CYP8B1 gene prevents the synthesis of cholate, which is a primary bile acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP8B1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYP8B1
Gene Accession No. O88962, Q9UNU6
Gene Alias 7 alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase; 7-alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase; 7-alpha-hydroxycholest-4-en-3-one 12-alpha-hydroxylase; CP8B; CYP12; Cyp8b1; CYPVIIIB1; cytochrome P450 8B1; cytochrome P450 8b1 sterol 12 alpha-hydrolase; cytochrome P450 family 8 subfamily B member 1; cytochrome P450, 8b1, sterol 12 alpha-hydrolase; cytochrome P450, family 8, subfamily B, polypeptide 1; cytochrome P450, subfamily VIIIB (sterol 12-alpha-hydroxylase), polypeptide 1; sterol 12-alpha-hydrolase; sterol 12-alpha-hydroxylase
Gene Symbols CYP8B1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYP8B1(Accession Q9UNU6), corresponding to amino acid residues G371-K421.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13124, 1582
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.