Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cypermethrin, 96%

Catalog No. AC456311000
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 52315-07-8 | C22H19Cl2NO3 | 416.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52315-07-8
Molecular Formula C22H19Cl2NO3
Molecular Weight (g/mol) 416.3
InChI Key KAATUXNTWXVJKI-UHFFFAOYSA-N
Synonym cypermethrin, barricade, ripcord, ustaad, ammo, supercypermethrin, cymbush, alpha-cypermethrin, cypermethrine, agrothrin
PubChem CID 2912
ChEBI CHEBI:4042
IUPAC Name [cyano-(3-phenoxyphenyl)methyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMILES CC1(C(C1C(=O)OC(C#N)C2=CC(=CC=C2)OC3=CC=CC=C3)C=C(Cl)Cl)C

Specifications

Density 1.1200g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Mixture of isomers) (HPLC)
Quantity 100 g
Specific Gravity 1.12
Solubility Information Solubility in water: insoluble.
Formula Weight 416.3
Percent Purity 96%
Chemical Name or Material Cypermethrin
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and
EINECSNumber 257-842-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.