Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyproconazole Monoclonal Antibody (2B10)
GREENER_CHOICE

Catalog No. PIMA549725
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549725 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549725 Supplier Invitrogen™ Supplier No. MA549725
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Cyproconazole is a fungicide that belongs to the triazole chemical class. It is used in agricultural applications to control fungal diseases on crops such as cereals, fruits, vegetables, and ornamental plants. Cyproconazole works by inhibiting the biosynthesis of ergosterol, an essential component of the fungal cell membrane. This disrupts the integrity of the cell membrane and ultimately leads to the death of the fungus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyproconazole
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 2B10
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of CCZ hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.