Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cyprodime hydrochloride
SDP

Catalog No. 260110
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-011-0 Supplier Tocris Bioscience™ Supplier No. 2601/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective μ antagonist

Selective μ-opioid receptor antagonist (Ki values are 5.4, 244.6 and 2187 nM for μ-, δ- and κ-opioid receptors respectively). Reduces levodopa-induced dyskinesia in the MPTP-lesioned primate model of Parkinson's disease.

Chemical Identifiers

CAS 2387505-50-0
Molecular Formula C22H30ClNO3

Specifications

Quantity 10 mg
Formula Weight 391.93
Purity >98%
Molecular Weight (g/mol) 391.94
Chemical Name or Material Cyprodime hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.