Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyproheptadine Hydrochloride, USP, 98.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601958 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-958 5 kg Poly Pail
18-601-955 5 g Amber Glass Bottle
18-601-956 25 g Amber Glass Bottle
18-601-959 100 g Amber Glass Bottle
18-601-960 500 g Amber Glass Bottle
18-601-957 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-601-958 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C19685KGBL
Add to Cart
Add to Cart
Request a Quote

C1968, 41354-29-4, C21H21N·HCl·11/2H2O

Spectrum™ Chemical Cyproheptadine Hydrochloride, USP , a first-generation antihistamine, is used to treat allergic reactions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 41354-29-4
Molecular Formula C42H50Cl2N2O3
Molecular Weight (g/mol) 701.77
MDL Number MFCD27967225,MFCD00012538,MFCD00012538,MFCD27967225,MFCD00242817
InChI Key ZEAUHIZSRUAMQG-UHFFFAOYSA-N
IUPAC Name dihydrogen bis(1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine) trihydrate dichloride
SMILES [H+].[H+].O.O.O.[Cl-].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12.CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

CAS Max % 100%
UN Number UN2811
Quantity 5 kg
Percent Purity 98.5 to 100.5%
Grade USP
Ignition Residue 0.1%
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.