Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cyproheptadine Monoclonal Antibody (3E10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549630
Description
Antibody tested in indirect ELISA.
Cyproheptadine is a medication that belongs to the group of first-generation antihistamines. It is commonly used to relieve symptoms of allergies, such as itching, sneezing, and runny nose. Cyproheptadine works by blocking the effects of histamine, a substance in the body that causes allergic symptoms. In addition to its antihistamine properties, cyproheptadine also has anticholinergic and sedative effects.
Specifications
Cyproheptadine | |
Monoclonal | |
3.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Mouse | |
Antibody | |
IgG2a |
ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
3E10 | |
Unconjugated | |
Eiproheptadine; Periactin; Periactinol | |
Conjugate of CHD hapten and KLH. | |
200 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction