Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyr61/CCN1 Antibody (3A7.1B8), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236490C
Description
Cyr61/CCN1 Monoclonal specifically detects Cyr61/CCN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cyr61/CCN1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1 | |
Mouse | |
Protein G purified | |
RUO | |
3491 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
3A7.1B8 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
O00622 | |
CCN1 | |
Partial recombinant protein from human Cyr61/CCN1 protein (between residues 100-300). [Swiss-Prot O00622] | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction