Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYRPTIC/CFC1 Antibody (MM0205-8D12) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | CYRPTIC/CFC1 |
|---|---|
| Clone | MM0205-8D12 |
| Concentration | LYOPH |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
CYRPTIC/CFC1 Monoclonal specifically detects CYRPTIC/CFC1 in Human samples. It is validated for Western Blot.Specifications
| CYRPTIC/CFC1 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| P0CG36 | |
| 55997 | |
| IgG1 | |
| Protein G purified |
| MM0205-8D12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213 | |
| CFC1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title