Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYRPTIC/CFC1 Antibody (MM0205-8D12), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212209FR
Description
CYRPTIC/CFC1 Monoclonal specifically detects CYRPTIC/CFC1 in Human samples. It is validated for Western Blot.Specifications
CYRPTIC/CFC1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CFC1 | |
Human recombinant CYRPTIC/CFC1 | |
0.1 mL | |
55997 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
MM0205-8D12 | |
Western Blot | |
CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213 | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction