Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYSLTR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102071 Supplier Invitrogen™ Supplier No. PA5102071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CLTR1.

The cysteinyl leukotrienes LTC4, LTD4, and LTE4 are important mediators of human bronchial asthma. Pharmacologic studies have determined that cysteinyl leukotrienes activate at least 2 receptors, the protein encoded by this gene and CYSLTR2. This encoded receptor is a member of the superfamily of G protein-coupled receptors. Activation of this receptor by LTD4 results in contraction and proliferation of smooth muscle, oedema, eosinophil migration and damage to the mucus layer in the lung.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYSLTR1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYSLTR1
Gene Accession No. Q9Y271
Gene Alias BB147369; CysLT 1; CysLT(1); Cyslt1; CysLT1 receptor; Cys-LT1 receptor; Cyslt1r; CYSLTR; Cysltr1; cysteinyl leukotriene 1 receptor short isoform; cysteinyl leukotriene D4 receptor; Cysteinyl leukotriene receptor 1; G-protein coupled receptor HG55; HG55; HMTMF81; LTD4 receptor; LTD4R; MGC46139
Gene Symbols CYSLTR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYSLTR1(Accession Q9Y271), corresponding to amino acid residues K139-L189.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10800
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.