Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYSLTR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528735
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528735 Supplier Invitrogen™ Supplier No. PA528735
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLaS3, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The cysteinyl leukotrienes LTC4, LTD4, and LTE4 are important mediators of human bronchial asthma. Pharmacologic studies have determined that cysteinyl leukotrienes activate at least 2 receptors, the protein encoded by this gene and CYSLTR2. This encoded receptor is a member of the superfamily of G protein-coupled receptors. Activation of this receptor by LTD4 results in contraction and proliferation of smooth muscle, oedema, eosinophil migration and damage to the mucus layer in the lung.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYSLTR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CYSLTR1
Gene Accession No. Q9Y271
Gene Alias BB147369; CysLT 1; CysLT(1); Cyslt1; CysLT1 receptor; Cys-LT1 receptor; Cyslt1r; CYSLTR; Cysltr1; cysteinyl leukotriene 1 receptor short isoform; cysteinyl leukotriene D4 receptor; Cysteinyl leukotriene receptor 1; G-protein coupled receptor HG55; HG55; HMTMF81; LTD4 receptor; LTD4R; MGC46139
Gene Symbols CYSLTR1
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of human CysLT 1 Receptor. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10800
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.