Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystathionase Antibody (OTI1E12), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270488UV
Description
Cystathionase Monoclonal specifically detects Cystathionase in Human, Mouse, Canine, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cystathionase | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CTH | |
Full length recombinant protein of human Cystathionase (NP_001893) produced in HEK293T cells. | |
0.1 mL | |
1491.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1E12 | |
Western Blot, Immunohistochemistry | |
cystathionase (cystathionine gamma-lyase), cystathionine gamma-lyase, cysteine desulfhydrase, EC 4.4.1, EC 4.4.1.1, gamma-cystathionase, homoserine deaminase, homoserine dehydratase, MGC9471 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Canine, Monkey, Primate | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction